3-(Difluoromethyl)aniline 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H34188-03
H34188-MD
CAAAH34188-03EA
470.84
CAD
CAAAH34188-03
CAAAH34188-MD
3-(Difluoromethyl)aniline 97%
3-(Difluoromethyl)aniline
Purity: 97%
CAS No.: 368-99-0
Molecular Formula: C7H7F2N
Formula Weight: 143.14
Boiling Point: 82°/4mm
UN No.: 2810
MDL No.: MFCD18384830
Formula:
C₇H₇F₂N MW: 143.137 g/mol Boiling Pt: 82 °C/4 mm Storage Temperature: Ambient |
MDL Number:
MFCD18384830 CAS Number: 368-99-0 UN: 2810 ADR: 6.1 III |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...