3-Amino-4-chloro-1H-indazole 97%

Supplier: Thermo Scientific Chemicals
Warning

H34143-03 H34143-06 H34143-MD
CAAAH34143-03EA 197.5 CAD
CAAAH34143-03 CAAAH34143-06 CAAAH34143-MD
3-Amino-4-chloro-1H-indazole 97%
3-Amino-4-chloro-1H-indazole

CAS: 20925-60-4 MDL: MFCD00663139

Formula: C₇H₆ClN₃
Melting Pt: 157…159 °C
Storage Temperature: Ambient
MDL Number: MFCD00663139
CAS Number: 20925-60-4

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR