10-(Pentafluorophenoxycarbonyl)decyltrimethoxysilane 95%

Supplier: Thermo Scientific Chemicals
Warning

H34117-03 H34117-MD
CAAAH34117-03EA 238.58 CAD
CAAAH34117-03 CAAAH34117-MD
10-(Pentafluorophenoxycarbonyl)decyltrimethoxysilane 95%
10-(Pentafluorophenoxycarbonyl)decyltrimethoxysilane

Purity: 95% CAS No.: 944721-52-2 Molecular Formula: C20H29F5O5Si Formula Weight: 472.52 Refractive Index: 1.4370-1.4410 Boiling Point: 145-150°/0.05mm MDL No.: MFCD13181915

Formula: C₂₀H₂₉F₅O₅Si
Boiling Pt: 145…150 °C/0.05 mm
Storage Temperature: Ambient
MDL Number: MFCD13181915
CAS Number: 944721-52-2

Order Now

Specification Test Results

Assay Minimum 95


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR