3-Amino-6-bromo-1H-indazole 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Bromo-1H-indazol-3-amine

H34095-03 H34095-09
CAAAH34095-03EA 80.11 CAD
CAAAH34095-03 CAAAH34095-09
3-Amino-6-bromo-1H-indazole 97%
3-Amino-6-bromo-1H-indazole

CAS: 404827-77-6 MDL: MFCD05665872

Formula: C₇H₆BrN₃
MW: 212.05 g/mol
Melting Pt: 227…230 °C
Storage Temperature: Ambient
MDL Number: MFCD05665872
CAS Number: 404827-77-6
UN: 2811
ADR: 6.1 III

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR