5-Methoxy-2-nitrophenylamine 98%

Supplier: Thermo Scientific Chemicals
Warning

H34046-03 H34046-09
CAAAH34046-03EA 32.55 CAD
CAAAH34046-03 CAAAH34046-09
5-Methoxy-2-nitrophenylamine 98%
5-Methoxy-2-nitrophenylamine

Purity: 98% CAS No.: 16133-49-6 Molecular Formula: C7H8N2O3 Formula Weight: 168.15 Melting Point: 125-127° MDL No.: MFCD00179573

Formula: C7H8N2O3
Melting Pt: 125…127 °C
Storage Temperature: Ambient
MDL Number: MFCD00179573
CAS Number: 16133-49-6
EINECS: 240-293-4

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR