1-Acetyl-5-indolinesulfonyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-acetylindoline-5-sulfonyl chloride, 1-acetyl-2,3-dihydro-1H-indole-5-sulfonyl chloride, 1-acetyl-2,3-dihydro-1H-indole-5-sulphonyl chloride, n-acetylindoline-5-sulfonyl chloride, n-acetylindoline-5-sulphonyl chloride, 1-acetylindoline-5-sulphonyl chloride

H34044-03 H34044-09
CAAAH34044-03EA 132.72 CAD
CAAAH34044-03 CAAAH34044-09
1-Acetyl-5-indolinesulfonyl chloride 97%
1-Acetyl-5-indolinesulfonyl chloride

Purity: 97% CAS No.: 52206-05-0 Molecular Formula: C10H10ClNO3S Formula Weight: 259.71 Melting Point: 172-174° UN No.: 3261 MDL No.: MFCD07368558

Formula: C₁₀H₁₀ClNO₃S
Melting Pt: 172…174 °C
Storage Temperature: Ambient
MDL Number: MFCD07368558
CAS Number: 52206-05-0
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR