10-(Pentafluorophenoxycarbonyl)decyltriethoxysilane 95%

Supplier: Thermo Scientific Chemicals
Warning

H34037-03 H34037-MD
CAAAH34037-03EA 489.8 CAD
CAAAH34037-03 CAAAH34037-MD
10-(Pentafluorophenoxycarbonyl)decyltriethoxysilane 95%
10-(Pentafluorophenoxycarbonyl)decyltriethoxysilane

Purity: 95% CAS No.: 1197981-08-0 Molecular Formula: C23H35F5O5Si Formula Weight: 514.61 Refractive Index: 1.4320-1.4360 Boiling Point: 155-160°/0.04mm MDL No.: MFCD13181916

Formula: C₂₃H₃₅F₅O₅Si
Boiling Pt: 155…160 °C/0.04 mm
Storage Temperature: Ambient
MDL Number: MFCD13181916
CAS Number: 1197981-08-0

Order Now

Specification Test Results

Assay Minimum 95


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR