12-Mercaptododecylphosphonic acid 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(12-Mercaptododecyl)phosphonic acid
H33969-03
H33969-MD
CAAAH33969-03EA
685.72
CAD
CAAAH33969-03
CAAAH33969-MD
12-Mercaptododecylphosphonic acid 95%
12-Mercaptododecylphosphonic acid
Purity: 95%
CAS No.: 159239-33-5
Molecular Formula: C12H27O3PS
Formula Weight: 282.38
Melting Point: 94-96°
MDL No.: MFCD11046116
Formula:
C₁₂H₂₇O₃PS MW: 282.379 g/mol Melting Pt: 94…96 °C Storage Temperature: Ambient |
MDL Number:
MFCD11046116 CAS Number: 159239-33-5 |
Specification Test Results
Assay Minimum | 95 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...