7-Bromo-5-fluorobenzo[b]furan 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H33948-03
H33948-MD
CAAAH33948-03EA
360.24
CAD
CAAAH33948-03
CAAAH33948-MD
7-Bromo-5-fluorobenzo[b]furan 97%
7-Bromo-5-fluorobenzo[b]furan
Purity: 97%
CAS No.: 253429-19-5
Molecular Formula: C8H4BrFO
Formula Weight: 215.02
MDL No.: MFCD09056780
Formula:
C₈H₄BrFO MW: 215.021 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09056780 CAS Number: 253429-19-5 |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...