3-Methoxythiophene-2-carboxaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

H33940-03 H33940-MD
CAAAH33940-03EA 350.76 CAD
CAAAH33940-03 CAAAH33940-MD
3-Methoxythiophene-2-carboxaldehyde 97%
3-Methoxythiophene-2-carboxaldehyde

Purity: 97% CAS No.: 35134-07-7 Molecular Formula: C6H6O2S Formula Weight: 142.18 Melting Point: 80-82° MDL No.: MFCD01571098

Formula: C₆H₆O₂S
Melting Pt: 80…82 °C
Storage Temperature: Ambient
MDL Number: MFCD01571098
CAS Number: 35134-07-7

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR