Diethyl-12-mercaptododecylphosphonate 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 12-Mercaptododecylphosphonic acid diethyl ester

H33935-03 H33935-MD
CAAAH33935-03EA 685.72 CAD
CAAAH33935-03 CAAAH33935-MD
Diethyl-12-mercaptododecylphosphonate 95%
Diethyl-12-mercaptododecylphosphonate

Purity: 95% CAS No.: 1049677-30-6 Molecular Formula: C16H35O3PS Formula Weight: 338.49 Refractive Index: 1.4610-1.4650 MDL No.: MFCD11046117

Formula: C₁₆H₃₅O₃PS
Storage Temperature: Ambient
MDL Number: MFCD11046117
CAS Number: 1049677-30-6

Order Now

Specification Test Results

Assay Minimum 95


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR