4-Phenoxybenzyl bromide 97%

Supplier: Thermo Scientific Chemicals
Danger

H33895-03 H33895-MD
CAAAH33895-03EA 260.7 CAD
CAAAH33895-03 CAAAH33895-MD
4-Phenoxybenzyl bromide 97%
4-Phenoxybenzyl bromide

CAS: 36881-42-2 MDL: MFCD04038693

Formula: C₁₃H₁₁BrO
Storage Temperature: Ambient
MDL Number: MFCD04038693
CAS Number: 36881-42-2
EINECS: 253-253-6
UN: 3265
ADR: 8,III

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR