Isopropylboronic acid pinacol ester 98%

Supplier: Thermo Scientific Chemicals
Warning

H33861-03 H33861-06 H33861-14
CAAAH33861-03EA 41.4 CAD
CAAAH33861-03 CAAAH33861-06 CAAAH33861-14
Isopropylboronic acid pinacol ester 98%
Isopropylboronic acid pinacol ester

CAS: 76347-13-2 MDL: MFCD05663856

Formula: C₉H₁₉BO₂
Boiling Pt: 115 °C/60 mm
Storage Temperature: Ambient
MDL Number: MFCD05663856
CAS Number: 76347-13-2

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR