Methyl-2-iodo-5-methylbenzoate 95%

Supplier: Thermo Scientific Chemicals
Warning

H33853-03 H33853-06
CAAAH33853-03EA 51.98 CAD
CAAAH33853-03 CAAAH33853-06
Methyl-2-iodo-5-methylbenzoate 95%
Methyl 2-iodo-5-methylbenzoate

CAS: 103440-52-4 MDL: MFCD12198129

Formula: C₉H₉IO₂
Storage Temperature: Ambient
MDL Number: MFCD12198129
CAS Number: 103440-52-4

Order Now

Specification Test Results

Assay Minimum 95

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR