3-Chloro-2-hydrazino-5-(trifluoromethyl)pyridine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H33850-03
H33850-06
CAAAH33850-03EA
118.5
CAD
CAAAH33850-03
CAAAH33850-06
3-Chloro-2-hydrazino-5-(trifluoromethyl)pyridine 97%
3-Chloro-2-hydrazino-5-(trifluoromethyl)pyridine
Purity: 97%
CAS No.: 89570-82-1
Molecular Formula: C6H5ClF3N3
Formula Weight: 211.60
Physical Form: Solid
Appearance: White to cream
Melting Point: 92-94°
UN No.: 2811
MDL No.: MFCD00067865
Formula:
C₆H₅ClF₃N₃ Melting Pt: 92…94 °C Storage Temperature: Ambient |
MDL Number:
MFCD00067865 CAS Number: 89570-82-1 UN: 2811 ADR: 6.1,III |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...