11-Aminoundecyltriethoxysilane 90
Supplier: Thermo Scientific Chemicals
|
Warning
|
H33835-03
H33835-MD
CAAAH33835-03EA
884.8
CAD
CAAAH33835-03
CAAAH33835-MD
11-Aminoundecyltriethoxysilane 90
11-Aminoundecyltriethoxysilane
Purity: 90+%
CAS No.: 116821-45-5
Molecular Formula: C17H39NO3Si
Formula Weight: 333.59
Boiling Point: 110-115°/0.02mm
MDL No.: MFCD08272792
Formula:
C₁₇H₃₉NO₃Si MW: 333.588 g/mol Boiling Pt: 110…115 °C/0.02 mm Storage Temperature: Ambient |
MDL Number:
MFCD08272792 CAS Number: 116821-45-5 |
Specification Test Results
Assay Minimum | 95 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...