Diethyl chlorophosphate 97+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H33820-14
H33820-22
CAAAH33820-14EA
67.31
CAD
CAAAH33820-14
CAAAH33820-22
Diethyl chlorophosphate 97+%
Diethyl chlorophosphate
CAS: 814-49-3
MDL: MFCD00009075
Formula:
C₄H₁₀ClO₃P Boiling Pt: 60 °C/2 mm Density: 1.194 Flash Pt: 61 °C (142 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009075 CAS Number: 814-49-3 EINECS: 212-369-4 UN: 2927 ADR: 6.1,II |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...