3-Fluoro-2-nitropyridine 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H33813-03
H33813-06
CAAAH33813-03EA
251.22
CAD
CAAAH33813-03
CAAAH33813-06
CAAAH33813-MD
3-Fluoro-2-nitropyridine 96%
3-Fluoro-2-nitropyridine
CAS: 54231-35-5
MDL: MFCD04114127
Formula:
C₅H₃FN₂O₂ Melting Pt: 30…35 °C Storage Temperature: Ambient |
MDL Number:
MFCD04114127 CAS Number: 54231-35-5 EINECS: 207-260-6 |
Specification Test Results
Assay Minimum | 96 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...