4-Amino-1-methyl-1H-pyrazole 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H33728-03
H33728-06
CAAAH33728-03EA
350.76
CAD
CAAAH33728-03
CAAAH33728-06
CAAAH33728-MD
4-Amino-1-methyl-1H-pyrazole 97%
4-Amino-1-methyl-1H-pyrazole
Purity: 97%
CAS No.: 69843-13-6
Molecular Formula: C4H7N3
Formula Weight: 97.12
Melting Point: 47-50°C
Beilstein Reference: 4368862
MDL No.: MFCD05667135
Formula:
C₄H₇N₃ Melting Pt: 47…50°C Storage Temperature: Refrigerator |
MDL Number:
MFCD05667135 CAS Number: 69843-13-6 |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...