1-[4-(4-Morpholinyl)phenyl]guanidine 98%

Supplier: Thermo Scientific Chemicals

H33726-03 H33726-06
CAAAH33726-03EA 118.82 CAD
CAAAH33726-03 CAAAH33726-06 CAAAH33726-14
1-[4-(4-Morpholinyl)phenyl]guanidine 98%
1-[4-(4-Morpholinyl)phenyl]guanidine

Purity: 98% CAS No.: 247234-41-9 Molecular Formula: C11H16NO2 Formula Weight: 220.27 Melting Point: ca 220°dec. MDL No.: MFCD11986905

Formula: C₁₁H₁₆N₄O
MW: 220.276 g/mol
Melting Pt: ca 220 °C (dec.)
Density: 1.309
Storage Temperature: Ambient
MDL Number: MFCD11986905
CAS Number: 247234-41-9

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR