6-Hydroxy-m-anisidine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Amino-4-methoxyphenol

H33724-03 H33724-06
CAAAH33724-03EA 129.88 CAD
CAAAH33724-03 CAAAH33724-06
6-Hydroxy-m-anisidine 97%
6-Hydroxy-m-anisidine

CAS: 20734-76-3 MDL: MFCD06616911

Formula: C₇H₉NO₂
Melting Pt: 135…137 °C
Storage Temperature: Ambient
MDL Number: MFCD06616911
CAS Number: 20734-76-3
EINECS: 687-284-5

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR