Benzomorpholine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Benzomorpholine, 3,4-Dihydro-2H-1,4-benzoxazine

H33641-03 H33641-06
CAAAH33641-03EA 186.44 CAD
CAAAH33641-03 CAAAH33641-06
Benzomorpholine 97%
Benzomorpholine

Purity: 97% CAS No.: 5735-53-5 Molecular Formula: C8H9NO Formula Weight: 135.17 Boiling Point: 85°/1mm MDL No.: MFCD02181098

Formula: C₈H₉NO
MW: 135.166 g/mol
Boiling Pt: 85 °C/1 mm
Storage Temperature: Ambient
MDL Number: MFCD02181098
CAS Number: 5735-53-5

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR