4-Acetyl-2-chloropyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(2-chloropyridin-4-yl)ethan-1-one, 1-(2-chloropyridine-4-yl)ethanone

H33619-03 H33619-06
CAAAH33619-03EA 180.12 CAD
CAAAH33619-03 CAAAH33619-06
4-Acetyl-2-chloropyridine 97%
4-Acetyl-2-chloropyridine

CAS: 23794-15-2 MDL: MFCD07699406

Formula: C₇H₆ClNO
MW: 155.58 g/mol
Melting Pt: 37…40 °C
Storage Temperature: Ambient
MDL Number: MFCD07699406
CAS Number: 23794-15-2
UN: 2811
ADR: 6.1 III

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR