4-Cyclohexylbenzenesulfonyl chloride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H33616-03
H33616-09
CAAAH33616-03EA
134.3
CAD
CAAAH33616-03
CAAAH33616-09
4-Cyclohexylbenzenesulfonyl chloride 97%
4-Cyclohexylbenzenesulfonyl chloride
Purity: 97%
CAS No.: 56354-57-5
Molecular Formula: C12H15ClO2S
Formula Weight: 258.77
Melting Point: 52-54°
UN No.: 3261
MDL No.: MFCD00625713
Formula:
C₁₂H₁₅ClO₂S Melting Pt: 52…54 °C Storage Temperature: Ambient |
MDL Number:
MFCD00625713 CAS Number: 56354-57-5 UN: 3261 ADR: "8",III |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...