4-Chloro-5-(2-thienyl)thieno[2,3-d]pyrimidine 97%
Supplier: Thermo Scientific Chemicals
H33610-03
H33610-06
CAAAH33610-03EA
149.15
CAD
CAAAH33610-03
CAAAH33610-06
CAAAH33610-MD
4-Chloro-5-(2-thienyl)thieno[2,3-d]pyrimidine 97%
4-Chloro-5-(2-thienyl)thieno[2,3-d]pyrimidine
Purity: 97%
CAS No.: 189681-04-7
Molecular Formula: C10H5ClN2S2
Formula Weight: 252.75
MDL No.: MFCD00174019
Formula:
C₁₀H₅ClN₂S₂ MW: 252.734 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00174019 CAS Number: 189681-04-7 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...