2-Phenoxybenzyl bromide 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H33585-03
H33585-06
H33585-MD
CAAAH33585-03EA
415.54
CAD
CAAAH33585-03
CAAAH33585-06
CAAAH33585-MD
2-Phenoxybenzyl bromide 97%
2-Phenoxybenzyl bromide
Purity: 97%
CAS No.: 82657-72-5
Molecular Formula: C13H11BrO
Formula Weight: 263.14
UN No.: 3265
MDL No.: MFCD01320513
Formula:
C₁₃H₁₁BrO MW: 263.134 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01320513 CAS Number: 82657-72-5 UN: 3265 ADR: "8",III |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...