Pyridazine-3-carbonitrile 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Cyanopyridazine

H33568-03 H33568-MD
CAAAH33568-03EA 350.76 CAD
CAAAH33568-03 CAAAH33568-MD
Pyridazine-3-carbonitrile 97%
Pyridazine-3-carbonitrile

Purity: 97% CAS No.: 53896-49-4 Molecular Formula: C5H3N3 Formula Weight: 105.10 Melting Point: 43-45° UN No.: 3439 MDL No.: MFCD09881239

Formula: C₅H₃N₃
Melting Pt: 43…45 °C
Storage Temperature: Ambient
MDL Number: MFCD09881239
CAS Number: 53896-49-4
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR