N-Formyl-L-leucine 90%, Technical Grade
Supplier: Thermo Scientific Chemicals
|
Warning
|
H33564-03
H33564-06
CAAAH33564-03EA
29.55
CAD
CAAAH33564-03
CAAAH33564-06
N-Formyl-L-leucine 90%, Technical Grade
N-Formyl-L-leucine
Purity: 90%
CAS No.: 6113-61-7
Molecular Formula: C7H13NO3
Formula Weight: 159.18
Storage Temperature: -30°C to -10°C
Melting Point: 142-145°
MDL No.: MFCD00055861
Formula:
C₇H₁₃NO₃ Melting Pt: 142…145 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00055861 CAS Number: 6113-61-7 EINECS: 228-080-4 |
Specification Test Results
Assay Minimum | 90 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...