11-Azidoundecyltriethoxysilane 95%
Supplier: Thermo Scientific Chemicals
H33524-03
H33524-MD
CAAAH33524-03EA
304.94
CAD
CAAAH33524-03
CAAAH33524-MD
11-Azidoundecyltriethoxysilane 95%
11-Azidoundecyltriethoxysilane
Purity: 95%
CAS No.: 663171-33-3
Molecular Formula: C17H37N3O3Si
Formula Weight: 359.58
Refractive Index: 1.4390-1.4430
Boiling Point: 110-115°/0.04mm
MDL No.: MFCD11982879
Formula:
C₁₇H₃₇N₃O₃Si |
MDL Number:
MFCD11982879 CAS Number: 663171-33-3 |
Specification Test Results
Assay Minimum | 95 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...