6-Chloro-2-methyl-4-pyrimidinol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Chloro-4-hydroxy-2-methylpyrimidine, 6-Chloro-2-methylpyrimidin-4(1H)-one

H33478-03 H33478-09
CAAAH33478-03EA 127.19 CAD
CAAAH33478-03 CAAAH33478-09
6-Chloro-2-methyl-4-pyrimidinol 97%
6-Chloro-2-methyl-4-pyrimidinol

Purity: 97% CAS No.: 17551-52-9 Molecular Formula: C5H5ClN2O Formula Weight: 144.56 Melting Point: 207-211° MDL No.: MFCD00030773

Formula: C₅H₅ClN₂O
MW: 144.558 g/mol
Melting Pt: 207…211 °C
Storage Temperature: Ambient
MDL Number: MFCD00030773
CAS Number: 17551-52-9

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR