4-{[3-(trifluoromethyl)pyridin-2-yl]oxy}benzenesulfonyl chloride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H33473-06
H33473-MD
CAAAH33473-03EA
172.36
CAD
CAAAH33473-03
CAAAH33473-06
CAAAH33473-MD
4-{[3-(trifluoromethyl)pyridin-2-yl]oxy}benzenesulfonyl chloride 97%
4-{[3-(trifluoromethyl)pyridin-2-yl]oxy}benzenesulfonyl chloride
Purity: 97%
CAS No.: 874839-13-1
Molecular Formula: C12H7ClF3NO3S
Formula Weight: 337.71
Melting Point: 63-65°
UN No.: 3261
MDL No.: MFCD07774164
Formula:
C₁₂H₇ClF₃NO₃S MW: 337.697 g/mol Melting Pt: 63…65 °C Storage Temperature: Ambient |
MDL Number:
MFCD07774164 CAS Number: 874839-13-1 UN: 3261 ADR: 8 III |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...