5-Amino-3-(4-chlorophenyl)-1,2,4-thiadiazole 96%

Supplier: Thermo Scientific Chemicals
Warning

H33467-03 H33467-MD
CAAAH33467-03EA 156.1 CAD
CAAAH33467-03 CAAAH33467-MD
5-Amino-3-(4-chlorophenyl)-1,2,4-thiadiazole 96%
5-Amino-3-(4-chlorophenyl)-1,2,4-thiadiazole

Purity: 96% CAS No.: 19922-07-7 Molecular Formula: C8H6ClN3S Formula Weight: 211.67 Flash Point: 110°(230°F) Melting Point: 183-185° MDL No.: MFCD07357326

Formula: C₈H₆ClN₃S
MW: 211.667 g/mol
Melting Pt: 183…185 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD07357326
CAS Number: 19922-07-7

Order Now

Specification Test Results

Assay Minimum 96


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR