4-Acetamidobenzyl alcohol 97%

Supplier: Thermo Scientific Chemicals
Warning

H33454-03 H33454-06
CAAAH33454-03EA 89.11 CAD
CAAAH33454-03 CAAAH33454-06
4-Acetamidobenzyl alcohol 97%
4-Acetamidobenzyl alcohol

CAS: 16375-88-5 MDL: MFCD00016868

Formula: C₉H₁₁NO₂
Melting Pt: 118…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00016868
CAS Number: 16375-88-5

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR