3-(2-Fluorophenyl)-1H-pyrazole 98%

Supplier: Thermo Scientific Chemicals
Danger

H33450-03 H33450-09
CAAAH33450-03EA 39.82 CAD
CAAAH33450-03 CAAAH33450-09
3-(2-Fluorophenyl)-1H-pyrazole 98%
3-(2-Fluorophenyl)-1H-pyrazole

Purity: 98% CAS No.: 149739-32-2 Molecular Formula: C9H7FN2 Formula Weight: 162.17 Melting Point: 58-61° MDL No.: MFCD00973917

Formula: C₉H₇FN₂
Melting Pt: 58…61 °C
Storage Temperature: Ambient
MDL Number: MFCD00973917
CAS Number: 149739-32-2

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR