4-Chloro-7-methyl-5,6,7,8-tetrahydrobenzo[b]thieno[2,3-d]pyrimidine 96%

Supplier: Thermo Scientific Chemicals
Warning

H33439-03 H33439-06
CAAAH33439-03EA 142.52 CAD
CAAAH33439-03 CAAAH33439-06
4-Chloro-7-methyl-5,6,7,8-tetrahydrobenzo[b]thieno[2,3-d]pyrimidine 96%
4-Chloro-7-methyl-5,6,7,8-tetrahydrobenzo[b]thieno[2,3-d]pyrimidine

Purity: 96% CAS No.: 137438-23-4 Molecular Formula: C11H11ClN2S Formula Weight: 238.73 Melting Point: 109-113° MDL No.: MFCD02333821

Formula: C₁₁H₁₁ClN₂S
MW: 238.733 g/mol
Melting Pt: 109…113 °C
Storage Temperature: Ambient
MDL Number: MFCD02333821
CAS Number: 137438-23-4

Order Now

Specification Test Results

Assay Minimum 96


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR