3-Amino-1H-indazole 97%

Supplier: Thermo Scientific Chemicals
Warning

H33435-03 H33435-09
CAAAH33435-03EA 125.14 CAD
CAAAH33435-03 CAAAH33435-09
3-Amino-1H-indazole 97%
3-Amino-1H-indazole

CAS: 874-05-5 MDL: MFCD00182045

Formula: C₇H₇N₃
Melting Pt: 150…152°C
Storage Temperature: Ambient
MDL Number: MFCD00182045
CAS Number: 874-05-5
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR