1-[4-(Trifluoromethyl)pyrimidin-2-yl]-1H-pyrazole-4-sulfonyl chloride 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H33424-03
H33424-MD
CAAAH33424-03EA
134.46
CAD
CAAAH33424-03
CAAAH33424-MD
1-[4-(Trifluoromethyl)pyrimidin-2-yl]-1H-pyrazole-4-sulfonyl chloride 95%
1-[4-(Trifluoromethyl)pyrimidin-2-yl]-1H-pyrazole-4-sulfonyl chloride
Purity: 95%
Molecular Formula: C8H4ClF3N4O2S
Formula Weight: 312.66
Melting Point: 97-99°
UN No.: 3261
MDL No.: MFCD11505058
Formula:
C₈H₄ClF₃N₄O₂S Melting Pt: 97…99 °C Storage Temperature: Ambient |
MDL Number:
MFCD11505058 CAS Number: 1215564-15-0 UN: 3261 ADR: "8",III |
Specification Test Results
Assay Minimum | 95 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...