2-(Trifluoromethyl)cyclohexanone 97%

Supplier: Thermo Scientific Chemicals
Warning

H33423-03 H33423-MD
CAAAH33423-03EA 205.4 CAD
CAAAH33423-03 CAAAH33423-MD
2-(Trifluoromethyl)cyclohexanone 97%
2-(Trifluoromethyl)cyclohexanone

CAS: 56734-74-8 MDL: MFCD00102125

Formula: C₇H₉F₃O
MW: 166.14 g/mol
Boiling Pt: 71…73/12 mm
Storage Temperature: Ambient
MDL Number: MFCD00102125
CAS Number: 56734-74-8
UN: 1224
ADR: 3 III

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR