Hydrogen chloride 3 mol/l in cyclopentyl methyl ether
Supplier: Thermo Scientific Chemicals
|
Danger
|
H33398-AE
H33398-AP
CAAAH33398-AEEA
109.34
CAD
CAAAH33398-AE
CAAAH33398-AP
Hydrogen chloride 3 mol/l in cyclopentyl methyl ether
Hydrogen chloride
CAS: 7647-01-0
MDL: MFCD00011324
Formula:
HCl Boiling Pt: 106 °C Density: 0.895 Flash Pt: 10 °C (50 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00011324 CAS Number: 7647-01-0 EINECS: 231-595-7 UN: 2924 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...