4-Bromomethyl-2'-fluorobiphenyl 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-fluoro-4'-bromomethylbiphenyl, 4'-(Bromomethyl)-2-fluoro-1,1'-biphenyl

H33396-03 H33396-MD
CAAAH33396-03EA 346.02 CAD
CAAAH33396-03 CAAAH33396-MD
4-Bromomethyl-2'-fluorobiphenyl 97%
4-Bromomethyl-2'-fluorobiphenyl

Purity: 97% Molecular Formula: C13H10BrF Formula Weight: 265.13 Melting Point: 72-74° MDL No.: MFCD15146811

Formula: C₁₃H₁₀BrF
MW: 265.13 g/mol
Melting Pt: 72…74 °C
Storage Temperature: Ambient
MDL Number: MFCD15146811
CAS Number: 193013-76-2

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR