1-Boc-4-(2-chloroethyl)piperazine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-(2-Chloroethyl)piperazine,N1-BOC protected, tert-Butyl 4-(2-chloroethyl)piperazine-1-carboxylate

H33349-03 H33349-06
CAAAH33349-03EA 219.62 CAD
CAAAH33349-03 CAAAH33349-06
1-Boc-4-(2-chloroethyl)piperazine 97%
1-Boc-4-(2-chloroethyl)piperazine

Purity: 97% CAS No.: 208167-83-3 Molecular Formula: C11H21ClN2O2 Formula Weight: 248.76 Melting Point: 63-65° MDL No.: MFCD08443993

Formula: C₁₁H₂₁ClN₂O₂
MW: 248.751 g/mol
Melting Pt: 63…65 °C
Storage Temperature: Ambient
MDL Number: MFCD08443993
CAS Number: 208167-83-3

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR