Methyl-2-hydroxynicotinate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methyl 2-hydroxynicotinate, 2-Hydroxynicotinic acid methyl ester
H33334-03
H33334-06
CAAAH33334-03EA
267.02
CAD
CAAAH33334-03
CAAAH33334-06
CAAAH33334-MD
Methyl-2-hydroxynicotinate 97%
Methyl-2-hydroxynicotinate
Purity: 97%
CAS No.: 10128-91-3
Molecular Formula: C7H7NO3
Formula Weight: 153.10
Physical Form: Solid
Appearance: Cream
Melting Point: 153-155°
MDL No.: MFCD00661282
Formula:
C₇H₇NO₃ Melting Pt: 153…155 °C Storage Temperature: Ambient |
MDL Number:
MFCD08275055 CAS Number: 67383-31-7 |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...