4-(5-Trifluoromethyl-2-pyridyloxy)benzenesulfonyl chloride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H33283-03
H33283-06
H33283-MD
CAAAH33283-03EA
143.46
CAD
CAAAH33283-03
CAAAH33283-06
CAAAH33283-MD
4-(5-Trifluoromethyl-2-pyridyloxy)benzenesulfonyl chloride 97%
4-(5-Trifluoromethyl-2-pyridyloxy)benzenesulfonyl chloride
Purity: 97%
CAS No.: 874838-96-7
Molecular Formula: C12H7ClF3NO3S
Formula Weight: 337.71
Melting Point: 51-53°
UN No.: 3261
MDL No.: MFCD07774162
Formula:
C₁₂H₇ClF₃NO₃S Melting Pt: 51…53 °C Storage Temperature: Ambient |
MDL Number:
MFCD07774162 CAS Number: 874838-96-7 UN: 3261 ADR: "8",III |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...