Palladium 5% on carbon, powder Type 490

Supplier: Thermo Scientific Chemicals
Warning

H33217-09 H33217-18
CAAAH33217-09EA 584.6 CAD
CAAAH33217-09 CAAAH33217-18
Palladium 5% on carbon, powder Type 490
Palladium

CAS No.: 7440-05-3 Molecular Formula: Pd UN No.: 1325 MDL No.: MFCD03457879

Formula: Pd
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6
UN: 1325
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR