4,4-Dimethyl-2-pyrrolidinone 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4,4-Dimethylpyrrolidin-2-one
H33209-03
H33209-MD
CAAAH33209-03EA
281.24
CAD
CAAAH33209-03
CAAAH33209-MD
4,4-Dimethyl-2-pyrrolidinone 95%
4,4-Dimethyl-2-pyrrolidinone
Purity: 95%
CAS No.: 66899-02-3
Molecular Formula: C6H11NO
Formula Weight: 113.20
Physical Form: Solid
Sensitivity: Air Sensitive
Appearance: White to cream
Melting Point: 38-40°
MDL No.: MFCD09864496
Formula:
C₆H₁₁NO MW: 113.16 g/mol Melting Pt: 38…40 °C Storage Temperature: Ambient |
MDL Number:
MFCD09864496 CAS Number: 66899-02-3 |
Specification Test Results
Assay Minimum | 95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...