4'-Fluorobiphenyl-4-carboxylic acid 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4'-Fluoro-[1,1'-biphenyl]-4-carboxylic acid, 4-(4-Fluorophenyl)benzoic acid

H33208-03 H33208-06
CAAAH33208-03EA 153.1 CAD
CAAAH33208-03 CAAAH33208-06
4'-Fluorobiphenyl-4-carboxylic acid 96%
4'-Fluorobiphenyl-4-carboxylic acid

CAS: 1399520 MDL: MFCD01631909

Formula: C₁₃H₉FO₂
MW: 216.21 g/mol
Melting Pt: 243…244 °C
Storage Temperature: Ambient
MDL Number: MFCD01631909
CAS Number: 5731-10-2

Order Now

Specification Test Results

Assay Minimum 96

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR