7-Bromo-1-methyl-3,4-dihydro-2(1H)-quinolinone 96%

Supplier: Thermo Scientific Chemicals
Warning

H33197-03 H33197-MD
CAAAH33197-03EA 410.8 CAD
CAAAH33197-03 CAAAH33197-MD
7-Bromo-1-methyl-3,4-dihydro-2(1H)-quinolinone 96%
7-Bromo-1-methyl-3,4-dihydro-2(1H)-quinolinone

Purity: 96% CAS No.: 1086386-20-0 Molecular Formula: C10H10BrNO Formula Weight: 240.10 Melting Point: 130-134° MDL No.: MFCD11655616

Formula: C₁₀H₁₀BrNO
Melting Pt: 130…134 °C
Storage Temperature: Ambient
MDL Number: MFCD11655616
CAS Number: 1086386-20-0

Order Now

Specification Test Results

Assay Minimum 96


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR