3-(Difluoromethyl)pyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

H33189-03 H33189-MD
CAAAH33189-03EA 175.38 CAD
CAAAH33189-03 CAAAH33189-MD
3-(Difluoromethyl)pyridine 97%
3-(Difluoromethyl)pyridine

CAS: 76541-44-1 MDL: MFCD11226584

Formula: C₆H₅F₂N
Boiling Pt: 42 °C/6 mm
Flash Pt: 54 °C (129 °F)
Storage Temperature: Ambient
MDL Number: MFCD11226584
CAS Number: 76541-44-1
EINECS: 278-487-6
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR