4-Phenoxybenzyl alcohol 97%

Supplier: Thermo Scientific Chemicals
Warning

H33138-03 H33138-06
CAAAH33138-03EA 290.72 CAD
CAAAH33138-03 CAAAH33138-06 CAAAH33138-MD
4-Phenoxybenzyl alcohol 97%
4-Phenoxybenzyl alcohol

Purity: 97% CAS No.: 2215-78-3 Molecular Formula: C13H12O2 Formula Weight: 200.24 MDL No.: MFCD01463970

Formula: C₁₃H₁₂O₂
Storage Temperature: Ambient
MDL Number: MFCD01463970
CAS Number: 2215-78-3

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR