2,1,3-Benzothiadiazole-5-carboxylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

H33095-03 H33095-06 H33095-MD
CAAAH33095-03EA 218.04 CAD
CAAAH33095-03 CAAAH33095-06 CAAAH33095-MD
2,1,3-Benzothiadiazole-5-carboxylic acid 97%
2,1,3-Benzothiadiazole-5-carboxylic acid

CAS: 16405-98-4 MDL: MFCD01647555

Formula: C₇H₄N₂O₂S
Melting Pt: 228…230 °C
Storage Temperature: Ambient
MDL Number: MFCD01647555
CAS Number: 16405-98-4

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR